Please login to your online account to display your discounted pricing

Phosphonoacetic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phosphonoacetic acid
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5O5P
Linear Formula HO2CCH2P(O)(OH)2
Formula Weight 140.03
MDL Number MFCD00004311
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC258291000

  SDS
thermo fisher scientific
AC258291000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC258290250

  SDS
thermo fisher scientific
AC258290250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 140.03
Quantity 100g
Color White
Infrared Spectrum Authentic
Molecular Formula C2H5O5P
CAS 4408-78-0
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00004311
Packaging Glass bottle
Linear Formula HO2CCH2P(O)(OH)2
Chemical Name or Material Phosphonoacetic acid
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.