Learn More
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals
$77.66 - $199.01
Chemical Identifiers
CAS | 940-71-6 |
---|---|
Molecular Formula | Cl6N3P3 |
Molecular Weight (g/mol) | 347.65 |
MDL Number | MFCD00006474 |
InChI Key | UBIJTWDKTYCPMQ-UHFFFAOYSA-N |
Synonym | phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer |
PubChem CID | 220225 |
IUPAC Name | 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
SMILES | N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC174280250
|
Thermo Scientific Chemicals
174280250 |
25 g | Glass bottle |
N/A
|
|
||||
AC174281000
|
Thermo Scientific Chemicals
174281000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
940-71-6 | |
347.65 | |
UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
220225 | |
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl |
Cl6N3P3 | |
MFCD00006474 | |
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
Specifications
111.0°C to 115.0°C | |
25 g | |
Cl6N3P3 | |
02,206; 04,386; 06,469 | |
Solubility in water: reacts. Other solubilities: soluble in ether, benzene, toluene | |
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl | |
347.65 | |
347.65 | |
Glass bottle | |
1.9800g/mL |
940-71-6 | |
98% | |
MFCD00006474 | |
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
220225 | |
98% | |
1.98 | |
Phosphonitrilic chloride trimer |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 213-376-8
RUO – Research Use Only