missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphomolybdic Acid Hydrate, TCI America™

Manufacturer:  TCI America P191025G

 View more versions of this product

Catalog No. P191025G



Specifications

Specifications

Phosphomolybdic Acid Hydrate
90°C
51429-74-4
MFCD00149913
Molybdo(VI)phosphoric Acid
Crystal-Powder at 20°C
Yellow
25g
H3Mo12O40P
3085
1825.48

Chemical Identifiers

51429-74-4
H3Mo12O40P
MFCD00149913
Molybdo(VI)phosphoric Acid
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0714

TSCA : Yes

Documents