Please login to your online account to display your discounted pricing

Phosphomolybdic acid hydrate, ACS reagent, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC417890250

 View more versions of this product

Catalog No. AC417890250

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Crystals
Formula Weight 1825.25
Quantity 25g
Color Yellow
Merck Index 14, 7342
Ammonium 100ppm max.
Heavy Metals (as Pb) 50ppm max.
Packaging Glass bottle
CAS 51429-74-4
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Additional Information Vapor Pressure: 4.0mmHg at 17.8°C
Chemical Name or Material Phosphomolybdic acid hydrate
Sulfate 250ppm max.
Molecular Formula H3Mo12O40P·xH2O
Linear Formula H3[P(Mo3O10)4]·xH2O
MDL Number MFCD00149913
Insoluble Matter 0.01% max.
Calcium (Ca) 200ppm max.
Iron (Fe) 50ppm max.
Chlorine (Cl) 200ppm max.
Synonym Molybdophosphoric acid
Health Hazard 1 Danger
CAS 12026-57-2

Specifications

Chemical Name or Material Phosphomolybdic acid hydrate
CAS 51429-74-4
CAS 12026-57-2
Purity Grade ACS Reagent Grade
Physical Form Crystals
Molecular Formula H3Mo12O40P·xH2O
Linear Formula H3[P(Mo3O10)4]·xH2O
Formula Weight 1825.25
MDL Number MFCD00149913
Synonym Molybdophosphoric acid
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

SDS & Safety Documents