Learn More
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 206381000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Phosphomolybdic acid hydrate | |
ca. 78% Phosphomolybdic Acid | |
Glass bottle | |
100 g | |
Crystals | |
MFCD00149913 | |
Molybdophosphoric acid | |
FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide | |
1825.25 |
Yellow | |
51429-74-4,12026-57-2 | |
47.5% min. (Mo) | |
H3Mo12O40P | |
H3[P(Mo3O10)4]·xH2O | |
14, 7342 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O | |
1825.40 | |
80% |
Chemical Identifiers
51429-74-4 | |
1825.40 | |
FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide |
H3Mo12O40P | |
MFCD00149913 | |
Molybdophosphoric acid | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO – Research Use Only