missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphomolybdic acid, ammonium salt hydrate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC317102500

 View more versions of this product

Catalog No. AC317102500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Powder
250g
H12Mo12N3O40P·xH2O
54723-94-3
MFCD00150423
200ppm max.
Ammonium phosphomolybdate hydrate
0.01% max.
500ppm max.
1876.35
Yellow
Phosphomolybdic acid, ammonium salt hydrate
(NH4)3PMo12O40·xH2O
50ppm max.
Plastic bottle
250ppm max.
50ppm max.

Chemical Identifiers

54723-94-3
(NH4)3PMo12O40·xH2O
MFCD00150423
Phosphomolybdic acid, ammonium salt hydrate
H12Mo12N3O40P·xH2O
1876.35
Ammonium phosphomolybdate hydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger