missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphomolybdic acid, 20 wt.% solution in ethanol, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368621000

 View more versions of this product

Catalog No. AC368621000


 

Specifications Description & Specifications

Specifications

1825.25
100mL
19 to 21%
13, 7428
H3Mo12O40P
Phosphomolybdic acid
Plastic bottle
20 wt% solution in ethanol
Solution
Clear yellow to green
0.9400g/mL
Molybdophosphoric acid
MFCD00011341
51429-74-4,64-17-5
H3[P(Mo3O10)4]

Chemical Identifiers

51429-74-4,64-17-5
H3[P(Mo3O10)4]
MFCD00011341
Phosphomolybdic acid, 20 wt% solution in ethanol
H3Mo12O40P
1825.25
Molybdophosphoric acid
19 to 21%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger