Please login to your online account to display your discounted pricing

Phospho-ZAP-70/SYK (Y319/Y352) Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: n3kobu5, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen Phospho-ZAP-70/SYK (Y319/Y352)
Clone n3kobu5
Host Species Mouse
Gene Alias Zeta chain-associated protein of 70 kD/Spleen tyrosine kinase
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123925

 
life technologies
LS46900641
25 tests Each for $201.55

501129064

 
life technologies
LS46900642
100 tests Each for $520.55
Description & Specifications

Specifications

Antigen Phospho-ZAP-70/SYK (Y319/Y352)
Clone n3kobu5
Host Species Mouse
Gene Alias Zeta chain-associated protein of 70 kD/Spleen tyrosine kinase
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

This n3kobu5 monoclonal antibody recognizes human and mouse zeta chain-associated protein of 70 kD (also known as ZAP-70) and spleen tyrosine kinase (also known as SYK) when phosphorylated on Y319 and Y352, respectively. ZAP-70 and SYK are members of the SYK protein tyrosine kinase (PTK) family that are phosphorylated and activated by Src family PTKs. ZAP-70/SYK Y319/Y352 are located in the so-called interdomain of ZAP-70/SYK (between the N-terminal dual SH2 domains and the C-terminal kinase domain).

Phosphorylation of ZAP-70 Y319 by Lck is necessary for T cell receptor (TCR)-dependent association of ZAP-70 with Lck and phospholipase C gamma and subsequent activation of calcium-dependent and Ras signaling cascades. SYK Y352 phosphorylation by Fyn/Lyn is critical for propagation of B cell receptor (BCR) signaling and for B cell development.

Specificity of this n3kobu5 clone was determined by ELISA, flow cytometry, and western blotting.