Please login to your online account to display your discounted pricing

Phospho-Tyrosine Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: pY20, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen Phospho-Tyrosine
Clone pY20
Host Species Murine
Gene Alias pTyr20
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123818

 
life technologies
LS46500141
25 Tests Each for $201.55

501123819

 
life technologies
LS46500142
100 Tests Each for $520.55
Description & Specifications

Specifications

Antigen Phospho-Tyrosine
Clone pY20
Host Species Murine
Gene Alias pTyr20
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

The pY20 monoclonal antibody recognizes phosphorylated tyrosine residues (p-Tyr) on proteins. Numerous intracellular signaling cascades are propagated via phosphorylation of specific tyrosine on signaling proteins. The detection of p-Tyr residues is valuable for the characterization and purification of phosphorylated proteins and the biochemical pathways that they are involved in.