Please login to your online account to display your discounted pricing

Phospho-Tyrosine Mouse anti-Human, Mouse, Clone: pY20, eBioscience™

Mouse Monoclonal Antibody

$56.55 - $143.55

Specifications

Antigen Phospho-Tyrosine
Clone pY20
Host Species Murine
Gene Alias pTyr20
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122685

 
life technologies
LS14500180
25μg Each for $56.55

501122686

 
life technologies
LS14500182
100μg Each for $143.55
Description & Specifications

Specifications

Antigen Phospho-Tyrosine
Clone pY20
Host Species Murine
Gene Alias pTyr20
Species Reactivity Human, Mouse
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The pY20 monoclonal antibody recognizes phosphorylated tyrosine residues (p-Tyr) on proteins. Numerous intracellular signaling cascades are propagated via phosphorylation of specific tyrosine on signaling proteins. The detection of p-Tyr residues is valuable for the characterization and purification of phosphorylated proteins and the biochemical pathways that they are involved in.