Please login to your online account to display your discounted pricing

Phospho-STAT5 (Y694) Mouse anti-Human, Mouse, FITC, Clone: SRBCZX, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $448.05

Specifications

Antigen Phospho-STAT5 (Y694)
Clone SRBCZX
Host Species Murine
Gene Alias signal transducer and activator of transcription 5, pSTAT5
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122165

 
life technologies
LS11901041
25 Tests Each for $187.05

501129150

 
life technologies
LS11901042
100 Tests Each for $448.05
Description & Specifications

Specifications

Antigen Phospho-STAT5 (Y694)
Clone SRBCZX
Host Species Murine
Gene Alias signal transducer and activator of transcription 5, pSTAT5
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

This SRBCZX monoclonal antibody recognizes signal transducer and activator of transcription 5 (STAT5) when phosphorylated on tyrosine 694. STAT proteins are activated by ligand binding to receptors, such as cytokine receptors, that associate with Janus kinase (JAK) family members. Following their phosphorylation by JAKs, STAT proteins translocate to the nucleus where they bind to DNA and regulate transcription of specific genes in a cell type- and cytokine-specific manner. In response to cytokines that signal through the common gamma chain such as IL-2, IL-7, and IL-15, STAT5 is phosphorylated on tyrosine 694 by JAK1 and JAK3. Cytokines such as IL-3, IL-5, and GM-CSF that signal via the common beta chain induce STAT5 phosphorylation on tyrosine 694 by JAK 2. Phosphorylation of STAT5 on tyrosine 694 is essential for STAT5 dimer formation, nuclear translocation, and DNA binding activity.

Specificity of this SRBCZX clone was determined by ELISA and flow cytometry.