Please login to your online account to display your discounted pricing

Phospho-SLP-76 (Y128) Mouse anti-Human, APC, Clone: HNDZ55, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $448.05

Specifications

Antigen Phospho-SLP-76 (Y128)
Clone HNDZ55
Host Species Mouse
Gene Alias Lymphocyte cytosolic protein 2, LCP2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123206

 
life technologies
LS17903741
25 tests Each for $187.05

501123207

 
life technologies
LS17903742
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen Phospho-SLP-76 (Y128)
Clone HNDZ55
Host Species Mouse
Gene Alias Lymphocyte cytosolic protein 2, LCP2
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This HNDZ55 monoclonal antibody recognizes human SH2 domain-containing leukocyte protein of 76 kD (SLP-76) when phosphorylated on tyrosine 128 (Y128). SLP-76 is phosphorylated by ZAP-70 downstream of the T cell receptor (TCR). SLP-76 contains an acidic region that includes a PEST domain, several tyrosine residues that are phosphorylated following TCR ligation, a proline-rich domain, and an SH2 domain. Numerous proteins associate with SLP-76 including Vav1, GADS, and PLC gamma 1, supporting the notion that SLP-76 functions as a critical adaptor protein downstream of the TCR. SLP-76-deficient T cell lines or mice deficient in SLP-76 have shown that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function.

This antibody recognizes only human SLP-76 when phosphorylated on Y128. It is not recommended for staining of mouse cells.