Please login to your online account to display your discounted pricing

Phospho-S6 Ribosomal Proteinl (S235/S236) Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: cupk43k, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen Phospho-S6 Ribosomal Proteinl (S235/S236)
Clone cupk43k
Host Species Murine
Gene Alias 40S ribosomal protein S6, Phosphoprotein NP33
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123926

 
life technologies
LS46900741
25 Tests Each for $201.55

501123927

 
life technologies
LS46900742
100 Tests Each for $506.05
Description & Specifications

Specifications

Antigen Phospho-S6 Ribosomal Proteinl (S235/S236)
Clone cupk43k
Host Species Murine
Gene Alias 40S ribosomal protein S6, Phosphoprotein NP33
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This cupk43k monoclonal antibody recognizes human and mouse ribosomal protein S6 (also known as 40S ribosomal protein S6, phosphoprotein NP33, RPS6, RS6, S6) when phosphorylated on serine 235 (S235, human) and serine 236 (S236, mouse). Ribosomal protein S6 is a component of the 40S subunit of the ribosome and is phosphorylated at multiple sites following stimulation of cells by growth factors, tumor promoting agents, or mitogens. Phosphorylation of ribosomal protein S6 by p70S6K and PKDCD results in upregulation of the translation of RNA coding for proteins involved in cell cycle entry. Ribosomal protein S6 is dephosphorylated upon growth arrest.

The specificity of the cupk43k monoclonal antibody was determined by western blotting.