missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-S6 Ribosomal Protein (S235/S236) Mouse anti-Human, Mouse, Clone: cupk43k, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $394.68

Specifications

Antigen Phospho-S6 Ribosomal Protein (S235/S236)
Clone cupk43k
Host Species Murine
Gene Alias 40S ribosomal protein S6, Phosphoprotein NP33
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122743 Life Technologies
LS14900780
25μg Each for $183.48
501122744 Life Technologies
LS14900782
100μg Each for $394.68
Specifications

Description

This cupk43k monoclonal antibody recognizes human and mouse ribosomal protein S6 (also known as 40S ribosomal protein S6, phosphoprotein NP33, RPS6, RS6, S6) when phosphorylated on serine 235 (S235, human) and serine 236 (S236, mouse). Ribosomal protein S6 is a component of the 40S subunit of the ribosome and is phosphorylated at multiple sites following stimulation of cells by growth factors, tumor promoting agents, or mitogens. Phosphorylation of ribosomal protein S6 by p70S6K and PKDCD results in upregulation of the translation of RNA coding for proteins involved in cell cycle entry. Ribosomal protein S6 is dephosphorylated upon growth arrest.

The specificity of the cupk43k monoclonal antibody was determined by western blotting.
Description & Specifications

Specifications

Phospho-S6 Ribosomal Protein (S235/S236)
Murine
Human, Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
cupk43k
40S ribosomal protein S6, Phosphoprotein NP33
Flow Cytometry (Intracellular Staining),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL