Please login to your online account to display your discounted pricing

Phospho-RSK (S380) Pan DuoSet IC, R&D Systems

$419.81 - $1,780.05

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DYC889BE

 
r & d systems
RDSDYC889BE
15 x 96-well plate Each for $1,780.05

DYC889B5

 
r & d systems
RDSDYC889B5
5 x 96-well plate Each for $828.14

DYC889B2

 
r & d systems
RDSDYC889B2
2 x 96-well plate Each for $419.81
Description & Specifications

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Sensitivity: 0pg/mL
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.