Please login to your online account to display your discounted pricing

Phospho-RSK (S380) Pan DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC889B2

 View more versions of this product

Catalog No. DYC889B2

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Format 2 x 96-well plate
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat