Please login to your online account to display your discounted pricing

Phospho-p70 S6 Kinase (T421/S424) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC89652

 View more versions of this product

Catalog No. DYC89652

  • / Each
 

Description & Specifications

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Gene ID (Entrez) 83840
Format Two 96-well plates
Species Human, Mouse, Rat
Quantity 1 Kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.