Please login to your online account to display your discounted pricing

Phospho-p38 gamma (T183/Y185) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC16642

 View more versions of this product

Catalog No. DYC16642

  • / Each
 

Description & Specifications

Specifications

Species Human, Mouse
Quantity 1 Kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gene ID (Entrez) 6300
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Packaging Quantity 2 Plates