Please login to your online account to display your discounted pricing

Phospho-p38 gamma (T183/Y185) DuoSet IC, R&D Systems

$419.81 - $419.81

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 6300
Species Human, Mouse
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC16642

 
r & d systems
RDSDYC16642
2 Plates Each for $419.81
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 6300
Species Human, Mouse

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.