missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-p38 alpha (T180/Y182) DuoSet IC, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

$428.87 - $1,818.46

Specifications

Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 1432
Species Human, Mouse, Rat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
DYC869B2 R & D Systems
RDSDYC869B2
Two 96-well plates Each for $428.87
DYC869B5 R & D Systems
RDSDYC869B5
Five 96-well plates Each for $846.01
DYC869BE R & D Systems
RDSDYC869BE
15 96-well plates Each for $1,818.46
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
1432
Human, Mouse, Rat