missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-NF kappa B p65 (S529) Mouse anti-Human, PE, Clone: B33B4WP, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $407.88

Specifications

Antigen Phospho-NF kappa B p65 (S529)
Clone B33B4WP
Host Species Murine
Gene Alias Nuclear factor kappa B p65 subunit, RelA
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122479 Life Technologies
LS12986341
25 Tests Each for $157.08
501129177 Life Technologies
LS12986342
100 Tests Each for $407.88
Specifications

Description

The B33B4WP monoclonal antibody recognizes human NF kappa B (NFkB) p65 subunit when serine 529 is phosphorylated. NFkB, also known as nuclear factor kappa-light chain enhancer of activated B cells, is a ubiquitous transcription factor that regulates the transcription of many genes involved in cell proliferation, apoptosis, development, immunity and cancer. Functional NFkB is a homo- or hetero-dimer composed of 5 members of the NFkB family: p65 (RelA), c-Rel, RelB, p50 (NFkB1, p105 precursor protein), and p52 (NFkB2, p100 precursor protein). The activity of the complex is negatively regulated by binding to IkB inhibitors that sequester NFkB into the cytoplasm, inhibiting its transcriptional activity. NFkB-activating agents like tumor necrosis factor (TNF) alpha, interleukin-1 beta, lipopolysaccharide, camptothecin, and phorbol ester (PMA) induce the phosphorylation and degradation of IkB, leading to the translocation of NFkB to the nucleus where it binds to kB motifs and regulates gene expression. The activity of p65-containing NFkB complexes is positively regulated by phosphorylation of the p65 subunit at serine 529 and/or serine 536.

The B33B4WP monoclonal antibody recognizes a single band at 65kDa in reduced SDS lysates from TNF alpha-stimulated HeLa cells.
Description & Specifications

Specifications

Phospho-NF kappa B p65 (S529)
Murine
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
B33B4WP
Nuclear factor kappa B p65 subunit, RelA
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer