Please login to your online account to display your discounted pricing

Phospho-mTOR (S2448) Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: MRRBY, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $530.70

Specifications

Antigen Phospho-mTOR (S2448)
Clone MRRBY
Host Species Murine
Gene Alias mammalian target of rapamycin, FRAP, RAFT
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123957

 
life technologies
LS46971841
25 Tests Each for $220.40

501123958

 
life technologies
LS46971842
100 Tests Each for $530.70
Description & Specifications

Specifications

Antigen Phospho-mTOR (S2448)
Clone MRRBY
Host Species Murine
Gene Alias mammalian target of rapamycin, FRAP, RAFT
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This MRRBY monoclonal antibody recognizes human and mouse mammalian target of rapamycin (also known as mTOR, FRAP, RAFT) when phosphorylated on S2448. mTOR is a serine/threonine protein kinase that functions as an ATP and amino acid sensor as well as to balance nutrient availability with cell growth, proliferation, motility, survival, protein synthesis, and transcription. Activated mTOR increases production of enzymes necessary for glycolysis and controls the uptake of glucose and other nutrients. Increased glucose uptake and metabolism helps fulfill the energy needs for mTOR-driven cell growth and proliferation. When sufficient nutrients are available, mTOR transmits a positive signal to p70 S6 kinase and participates in the inactivation of the eIF4E inhibitor, 4E-BP1. mTOR is phosphorylated at S2448 via the PI3 kinase/Akt signaling pathway and is autophosphorylated at Ser2481. Due to its critical role in regulation of cell growth, survival, and metabolism, and because it is often abnormally regulated in tumors, mTOR is under investigation as a potential target for anti-cancer therapy.