Please login to your online account to display your discounted pricing

Phospho-Mcl-1 (S159) Mouse anti-Human, Mouse, APC, Clone: RBCERNR, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $491.55

Specifications

Antigen Phospho-Mcl-1 (S159)
Clone RBCERNR
Host Species Mouse
Gene Alias BCL2L3, EAT
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123208

 
life technologies
LS17903841
25 tests Each for $201.55

501123209

 
life technologies
LS17903842
100 tests Each for $491.55
Description & Specifications

Specifications

Antigen Phospho-Mcl-1 (S159)
Clone RBCERNR
Host Species Mouse
Gene Alias BCL2L3, EAT
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This RBCERNR monoclonal antibody recognizes human and mouse myeloid cell leukemia sequence 1 (Mcl-1) when phosphorylated on serine 159 (S159). Mcl-1 is an anti-apoptotic protein that is a member of the Bcl-2 family of proteins important for regulation of cell survival/apoptosis. Mcl-1 is primarily localized to the outer membrane of mitochondria where it prevents cytochrome c release via dimerization with other Bcl-2 family members such as Bim. PI3K activation of AKT results in the phosphorylation of GSK3 beta at serine 9 (S9) resulting in destabilization and degradation of GSK3 beta. Loss of GSK3 beta prevents phosphorylation of Mcl-1 on S159 and its subsequent ubiquitnation and degradation. Mice conditionally lacking Mcl-1 in lymphocytes showed that Mcl-1 is essential during early lymphoid development and for the maintenance of mature lymphocytes.