missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-LCK (Y505) Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: SRRCHA, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen Phospho-LCK (Y505)
Clone SRRCHA
Host Species Murine
Gene Alias lymphocyte-specific protein tyrosine kinase, Leukocyte C-terminal Src kinase, p56lck
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123935 Life Technologies
LS46907641
25 Tests Each for $201.55
501123936 Life Technologies
LS46907642
100 Tests Each for $520.55
Specifications

Description

This SRRCHA monoclonal antibody recognizes human and mouse LCK when phosphorylated on tyrosine 505 (Y505). Y505 is a negative-regulatory site that is phosphorylated by the kinase CSK. LCK is expressed in T cells where it associates with the cytoplasmic tails of the CD4 and CD8 co-receptors on T helper cells and cytotoxic T cells, respectively. When the TCR is engaged, LCK acts to phosphorylate the intracellular chains of CD3 and zeta chains of the TCR complex, allowing the cytoplasmic tyrosine kinase, ZAP-70, to bind to them. LCK then phosphorylates and activates ZAP-70, which then phosphorylates the adaptor protein, LAT, that serves as a docking site for a number of other proteins, including SLP-76, Grb2, and phospholipase C gamma1. Mice lacking expression of LCK show defects in T cell development and function.
Description & Specifications

Specifications

Phospho-LCK (Y505)
Murine
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SRRCHA
lymphocyte-specific protein tyrosine kinase, Leukocyte C-terminal Src kinase, p56lck
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.06μg)/test