missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-H2A.x (S139) Mouse anti-Human, Mouse, PE, Clone: CR55T33, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen Phospho-H2A.x (S139)
Clone CR55T33
Host Species Murine
Gene Alias Histone H2AX
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122481 Life Technologies
LS12986542
100 Tests Each for $433.55
501122480 Life Technologies
LS12986541
25 Tests Each for $187.05
Specifications

Description

The CR55T33 monoclonal antibody recognizes phosphorylated serine 139 of human and mouse H2AX. H2AX is a member of the H2A histone family that complex with DNA and other histones to form the repeating nucleosome units characteristic of eukaryotic chromatin. Nucleosomes consist of approximately 147 base pairs of DNA wrapped around an octamer of histones composed of two each of the four histone proteins: H2A, H2B, H3 and H4. After induction of DNA damage such as double-strand breaks by irradiation, genotoxic stresses, replication errors or gene recombination, PI3K-like kinases (e.g., ataxia telangiectasia mutated (ATM), ataxia telangiectasia Rad-3-related (ATR), and DNA-dependent protein kinase (DNA-PK) are activated to phosphorylate serine 139 in H2AX. This early phosphorylation event plays a critical role in recruiting proteins involved in DNA repair.

The monoclonal antibody CR55T33 recognizes a single band of approximately 15kDa on reduced cell lysates from Jurkat cells stimulated with etoposide.
Description & Specifications

Specifications

Phospho-H2A.x (S139)
Murine
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CR55T33
Histone H2AX
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test