Please login to your online account to display your discounted pricing

Phospho-GSK-3 beta (S9) DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC15905

 View more versions of this product

Catalog No. DYC15905

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2932
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Format Five 96-well plates
Species Human, Mouse, Rat
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.