Please login to your online account to display your discounted pricing

Phospho-GSK-3 beta (S9) DuoSet IC, R&D Systems

$419.81 - $828.14

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Gene ID (Entrez) Format Price Quantity    

DYC15905

 
r & d systems
RDSDYC15905
2932 Five 96-well plates Each for $828.14

DYC15902

 
r & d systems
RDSDYC15902
2932 Two 96-well plates Each for $419.81
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Quantity 1 Kit
Species Human, Mouse, Rat

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.