missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-GSK-3 alpha/beta (S21/S9) DuoSet IC, R&D Systems

$428.87 - $846.01

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
DYC26302 R & D Systems
RDSDYC26302
2 x 96-well plate Each for $428.87
DYC26305 R & D Systems
RDSDYC26305
5 x 96-well plate Each for $846.01
Specifications

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

ELISA
ELISA kit
Human, mouse, rat
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit