missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC, R&D Systems

$428.87 - $1,818.46

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
DYC1018B2 R & D Systems
RDSDYC1018B2
2 x 96-well plate Each for $428.87
DYC1018BE R & D Systems
RDSDYC1018BE
15 x 96-well plate Each for $1,818.46
DYC1018B5 R & D Systems
RDSDYC1018B5
5 x 96-well plate Each for $846.01
Specifications

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

ELISA
ELISA kit
Human, mouse, rat
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit