missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-ERK1/2 (T202/Y204) Mouse anti-Human, Mouse, PE-eFluor 610, Clone: MILAN8R, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $447.48

Specifications

Antigen Phospho-ERK1/2 (T202/Y204)
Clone MILAN8R
Host Species Murine
Gene Alias MAPK, ERK, p42/p44
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124883 Life Technologies
LS61910941
25 Tests Each for $183.48
501129114 Life Technologies
LS61910942
100 Tests Each for $447.48
Specifications

Description

This MILAN8R monoclonal antibody recognizes human and mouse extracellular signal-regulated kinases 1 and 2 (also known as ERK1/2, p44/p42, or MAPK3/1) when phosphorylated on T202/Y204. ERK1/2 belong to a family of conserved serine/threonine protein kinases known as mitogen-activated protein kinases (MAPKs) that are involved in many cellular programs such as proliferation, differentiation, motility, and survival. ERK1/2 signaling is activated in response to numerous extracellular stimuli including mitogens, growth factors, and cytokines. The primary activators of ERK1/2 are MEK1 and MEK2 which act by phosphorylating the activation loop residues T202/Y204 and T185/Y187 in ERK1 and ERK2, respectively. Several downstream targets of ERK1/2 have been identified, including p90RSK and the transcription factor Elk-1. ERK1/2 are negatively regulated by MAPK phosphatases, known as DUSPs or MKPs, as well as by chemical inhibitors of MEK including U0126 and PD98059. Disruption of the ERK pathway is common in many types of cancer.

Specificity of this MILAN8R clone was determined by ELISA, flow cytometry, and western blotting.
Description & Specifications

Specifications

Phospho-ERK1/2 (T202/Y204)
Murine
Flow Cytometry (Intracellular Staining)
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test
MILAN8R
MAPK, ERK, p42/p44
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer