missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) InstantOne™ ELISA Kit

Specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 in cell lysates. Detection of rat ERK 1/2 is expected. The InstantOne™ ELISA kit allows for analysis of samples in approximately one hour.

$567.24 - $9,032.05

Specifications

For Use With (Application) ELISA
Synonym p44/p42, MAP Kinase 1/2
Target ERK 1, ERK 2
Storage Requirements 2° to 8°C
Species Hamster, Human, Mouse
View More Specs

Alert:

Please contact Technical Service for a lot specific COA

Disclaimers:

For Research Use Only


Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Capture Antibody Reagent
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate seal


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125021 Life Technologies
LS8586012103
10 x 384 tests Each for $9,032.05
501125022 Life Technologies
LS8586012253
25 x 384 tests N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
501125023 Life Technologies
LS8586012503
50 x 384 tests N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
50246231 Life Technologies
LS858601211
96 Tests Each for $567.24
Specifications

Description

All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Description & Specifications

Specifications

ELISA
ERK 1, ERK 2
Hamster, Human, Mouse
p44/p42, MAP Kinase 1/2
2° to 8°C