Invitrogen™ eBioscience™ phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) InstantOne™ ELISA Kit
Specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 in cell lysates. Detection of rat ERK 1/2 is expected. The InstantOne™ ELISA kit allows for analysis of samples in approximately one hour.
Manufacturer:  Invitrogen™ LS8586012503
 View more versions of this product
Catalog No. 501125023
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Please contact Technical Service for a lot specific COA
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Capture Antibody Reagent
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate seal
Description
Description
All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Specifications
Specifications
ELISA
Hamster, Human, Mouse
ERK 1, ERK 2
50 x 384 tests
2° to 8°C
p44/p42, MAP Kinase 1/2