Please login to your online account to display your discounted pricing

Phospho-BTK/ITK (Y551/Y511) Mouse anti-Human, Mouse, Clone: M4G3LN, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen Phospho-BTK/ITK (Y551/Y511)
Clone M4G3LN
Host Species Mouse
Gene Alias Bruton’s tyrosine kinase (Btk), agammaglobulinaemia tyrosine kinase, B-cell progenitor kinase/IL-2 inducible T-cell kinase (Itk), T-cell-specific kinase, Tyrosine-protein kinase Lyk
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122747

 
life technologies
LS14901580
25μg Each for $216.05

501129061

 
life technologies
LS14901582
100μg Each for $506.05
Description & Specifications

Specifications

Antigen Phospho-BTK/ITK (Y551/Y511)
Clone M4G3LN
Host Species Mouse
Gene Alias Bruton’s tyrosine kinase (Btk), agammaglobulinaemia tyrosine kinase, B-cell progenitor kinase/IL-2 inducible T-cell kinase (Itk), T-cell-specific kinase, Tyrosine-protein kinase Lyk
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This M4G3LN monoclonal antibody recognizes human and mouse Bruton’s tyrosine kinase (also known as BTK, agammaglobulinaemia tyrosine kinase, B-cell progenitor kinase)/interleukin-2-inducible tyrosine kinase (also known as ITK, T-cell-specific kinase, Tyrosine-protein kinase Lyk) when phosphorylated on Y551 and Y511, respectively.

In addition to Src family and SYK family kinases, B cells and T cells express a third family of receptor-proximal kinases called the Tec kinase family. The B cell-specific Tec family kinase is BTK; T cells express ITK as well as Tec and Rlk. Tec family kinases play an indispensable role in activation of phospholipase C and activation of calcium-dependent signaling cascades following antigen receptor triggering. BTK has been implicated in propagating signals downstream of LPS-TLR4 in B cells via signaling through the TLR4 co-receptor RP105/CD180. Mice congenitally lacking BTK or ITK show profound defects in B cell or T cell development and signaling, respectively. People with mutations or deletions in BTK suffer from X-linked agammaglobulinemia and X-linked immunodeficiency.

Specificity of this M4G3LN clone was determined by ELISA, flow cytometry, and western blotting.