Please login to your online account to display your discounted pricing

Phospho-Akt (S473) Pan DuoSet IC, R&D Systems

$428.87 - $1,818.46

Specifications

Applications Development of sandwich ELISA to measure phosphorylated human/mouse/rat Akt in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DYC887BE

 
r & d systems
RDSDYC887BE
15 x 96-well plate Each for $1,818.46

DYC887B2

 
r & d systems
RDSDYC887B2
2 x 96-well plate Each for $428.87

DYC887B5

 
r & d systems
RDSDYC887B5
5 x 96-well plate Each for $846.01
Description & Specifications

Specifications

Applications Development of sandwich ELISA to measure phosphorylated human/mouse/rat Akt in cell lysates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Human, mouse, rat

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.