Invitrogen™ eBioscience™ phospho-AKT 1/2/3 (Ser473) InstantOne™ ELISA Kit
Specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster AKT 1/2/3 in cell lysates. Detection of rat AKT 1/2/3 is expected. The InstantOne™ ELISA kit allows for analysis of samples in approximately one hour.
Manufacturer:  Invitrogen™ LS8586042503
 View more versions of this product
Catalog No. 501125026
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Please contact Technical Service for a lot specific COA
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-AKT 1/2/3 (Ser473) Capture Antibody Reagent
phospho-AKT 1/2/3 (Ser473) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate seal
Description
Description
All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Specifications
Specifications
ELISA
Hamster, Human, Mouse
AKT
50 x 384 tests
2° to 8°C
PKB, Protein Kinase B, RAC-PK-alpha
Documents