Please login to your online account to display your discounted pricing

Phospho-4E-BP1 (T36/T45) Mouse anti-Human, Mouse, PE, Clone: V3NTY24, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen Phospho-4E-BP1 (T36/T45)
Clone V3NTY24
Host Species Murine
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122419

 
life technologies
LS12910741
25 Tests Each for $201.55

501122420

 
life technologies
LS12910742
100 Tests Each for $506.05
Description & Specifications

Specifications

Antigen Phospho-4E-BP1 (T36/T45)
Clone V3NTY24
Host Species Murine
Species Reactivity Human, Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The V3NTY24 monoclonal antibody recognizes human and mouse eukaryotic translation initiation factor eIF4E-binding protein 1 (4E-BP1) when phosphorylated at threonine 37 and/or threonine 46. 4E-BP1 is a member of a family of translation repressor proteins that include 4E-BP2 and 4E-BP3. In its non-phosphorylated form, 4E-BP1 binds to the eIF4E translation initiation factor and represses cap-dependent translation. Phosphorylation of 4E-BP1 at multiple sites is necessary to disrupt this interaction and de-repress cap-dependent translation. Studies have identified several kinases that phosphorylate 4E-BP1. For instance, FRAP/mTOR phosphorylates Thr37 and Thr46, while ATM phosphorylates Ser111. Phosphorylation of 4E-BP1 at Thr37 and Thr46 is inhibited by the PI3 kinase inhibitors LY294002 and wortmannin.