Please login to your online account to display your discounted pricing

Phosphate Standard, (1mL = 0.5mg PO4), Certified, 500ppm ±5ppm, LabChem™

$88.74 - $88.74

Chemical Identifiers

Chemical Name or Material Phosphate Standard
CAS 7732-18-5
CAS Max % 99.93
CAS 7778-77-0
CAS Max % 0.07
Name Note (1mL = 0.5mg PO4)
Purity Grade Certified Grade
Linear Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC185801

 
lab chem
LC185801
Poly Bottle 500mL Each for $88.74
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 136.09
Chemical Name or Material Phosphate Standard
Color Clear, colorless
Quantity 500mL
Name Note (1mL = 0.5mg PO4)
Physical Form Liquid
Assay Percent Notes as PO4
Specific Gravity 1g/cm3
Concentration 500ppm ±5ppm
Linear Formula KH2PO4
CAS 7732-18-5
Decomposition Information Phosphorus oxides
Identification Passes Test
CAS Max % 99.93
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7778-77-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.07
Recommended Storage Room Temperature
Density 1g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature