Please login to your online account to display your discounted pricing

PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents


Commonly used buffer

Quantity 4L
Packaging Poly CUBE

Manufacturer: fisher scientific company  BP3994

 View more versions of this product

Catalog No. BP3994

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4L
DNase DNase free
Protease Protease free
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.119M Phosphate Buffer
Chloride 0.136 to 0.144M
Conductivity 14000 to 17800μmhos/cm
Synonym PBS
CAS 7647-14-5
Name Note 10X Solution
CAS 7558-79-4
Recommended Storage RT
Chemical Name or Material Phosphate Buffered Saline
CAS 7447-40-7
Packaging Poly CUBE
CAS 7778-77-0
Health Hazard 2 CAUTION!
CAS 7732-18-5
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Avoid contact with skin, eyes and clothing. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Phosphate Buffered Saline
Name Note 10X Solution
CAS 7732-18-5
CAS 7647-14-5
CAS 7447-40-7
CAS 7558-79-4
CAS 7778-77-0
Physical Form Liquid
Synonym PBS

  •  Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
  • pH of a 1X solution made from this concentrated solution is ~7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Avoid contact with skin, eyes and clothing. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

RT

SDS & Safety Documents