Alfa Aesar™ Phosphate-Buffered Saline (10X), pH 7.4, low endotoxin
Manufacturer:  Alfa Aesar™ J67512AE
 View more versions of this product
Catalog No. AAJ67512AE
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Contains 100mM sodium phosphate dibasic, 20mM potassium phosphate monobasic, 27mM KCl, 1.37M NaCl, pH 7.4 ± 0.2.
Colorless
100mL
Liquid
Phosphate-Buffered Saline