Please login to your online account to display your discounted pricing

Phosphate Buffer pH7 For Hematology, EMD Millipore


Quantity 4L
Packaging Poly Bottle

Manufacturer: emd millipore corporation  121985

 View more versions of this product

Catalog No. M121985

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4L
Physical Form Liquid
Name Note For Hematology
Packaging Poly Bottle
Chemical Name or Material Phosphate Buffer pH7

Specifications

Chemical Name or Material Phosphate Buffer pH7
Name Note For Hematology
Physical Form Liquid