missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphate Buffer pH7 For Hematology, EMD Millipore

$114.54 - $308.00

Chemical Identifiers

Physical Form Liquid
Chemical Name or Material Phosphate Buffer pH7, For Hematology
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M121975 Emd Millipore Corporation
121975
Poly Bottle 1L Each for $114.54
M121985 Emd Millipore Corporation
121985
Poly Bottle 4L Each for $308.00
Specifications Description & Specifications

Chemical Identifiers

Liquid
Phosphate Buffer pH7, For Hematology

Specifications

Liquid
Poly Bottle
1L
Phosphate Buffer pH7, For Hematology