Please login to your online account to display your discounted pricing

Phosphate Buffer pH 6.8, for Phenols, LabChem™

$59.31 - $59.31

Chemical Identifiers

Chemical Name or Material Phosphate Buffer
CAS 7732-18-5
CAS Max % 82.23
CAS 7758-11-4
CAS Max % 10.54
CAS 7778-77-0
CAS Max % 7.23
Name Note For Phenols
Purity Grade Certified Grade
Linear Formula KH2PO4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC185601

 
lab chem
LC185601
500mL Each for $59.31
Description & Specifications

Specifications

pH 6.8 ± 0.1
Purity Grade Certified Grade
Formula Weight 136.09
Chemical Name or Material Phosphate Buffer
Traceability to NIST Traceable to NIST
CAS Max % 7.23
Decomposition Information Gaseous ammonia; Phosphorus oxides
Color clear, colorless solution
Quantity 500mL
Name Note For Phenols
Physical Form Liquid
Linear Formula KH2PO4
CAS 7732-18-5
CAS Max % 82.23
Identification Passes Test
CAS 7758-11-4
Packaging Poly Bottle
CAS Max % 10.54
Solubility Soluble in water
CAS 7778-77-0
Storage Information Room Temperature