Please login to your online account to display your discounted pricing

Phosphate Buffer, 0.05M, Fisher Chemical


Quantity 1L
Packaging Poly Bottle

Manufacturer: ricca chemical company  SB96051

 View more versions of this product

Catalog No. SB9605-1

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1L
Chemical Name or Material Phosphate Buffer, 0.05M
Methods For AOAC Method 968.13
ChemAlert Storage Symbol Gray
CAS 7732-18-5
CAS 6153-56-6
CAS Min % 91.0
CAS Min % 0.18
CAS 7778-77-0
CAS Max % 0.27
CAS Max % 0.19
CAS Max % 0.09
CAS Min % 0.26
CAS 7664-38-2
CAS Max % 5.99
CAS 64-19-7
CAS Min % 0.09
CAS Max % 95.0
Packaging Poly Bottle
CAS 79-09-4
CAS Min % 0.31
CAS Max % 0.32
CAS Min % 5.76

Specifications

Chemical Name or Material Phosphate Buffer, 0.05M
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 64-19-7
CAS Min % 5.76
CAS Max % 5.99
CAS 7778-77-0
CAS Min % 0.31
CAS Max % 0.32
CAS 7664-38-2
CAS Min % 0.26
CAS Max % 0.27
CAS 6153-56-6
CAS Min % 0.18
CAS Max % 0.19
CAS 79-09-4
CAS Min % 0.09
CAS Max % 0.09
Accessories