missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphate (as Phosphorus) Standard, (1mL = 1mg P), Certified, 1000ppm ±10ppm, LabChem™

Manufacturer:  Lab Chem LC185901

Catalog No. LC185901



Description

Description

Specifications

Specifications

Phosphate (as Phosphorus) Standard
7778-77-0,7732-18-5
as P
KH2PO4
1000ppm ±10ppm
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
Liquid
Passes Test
1g/cm3
500mL
(1mL = 1mg P)
99.56,0.44
H2KO4P
MFCD00011401,MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
516951
136.09
Certified
Poly Bottle
Colorless
Soluble in water

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
CHEBI:63036
[K+].OP(O)([O-])=O
H2KO4P
MFCD00011401,MFCD00147253
516951
potassium dihydrogen phosphate
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Product Certifications
Promotions

Promotions