Please login to your online account to display your discounted pricing

Phosphate (as Phosphorus) Standard, (1mL = 0.05mg P), Certified, 50ppm ±1ppm, LabChem™

$36.89 - $53.54

Chemical Identifiers

Chemical Name or Material Phosphate (as Phosphorus) Standard
CAS 7732-18-5
CAS Max % 99.98
CAS 7778-77-0
CAS Max % 0.02
Name Note (1mL = 0.05mg P)
Purity Grade Certified Grade
Linear Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC186001

 
lab chem
LC186001
Poly Bottle 500mL Each for $53.54

LC186007

 
lab chem
LC186007
Poly Bottle 125mL Each for $36.89
Description & Specifications

Specifications

Formula Weight 136.09
Purity Grade Certified Grade
Physical Form Liquid
Packaging Poly Bottle
Color Clear, colorless
Quantity 500mL
CAS 7778-77-0
CAS 7732-18-5
Name Note (1mL = 0.05mg P)
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
CAS Max % 0.02
Recommended Storage Room Temperature
Linear Formula KH2PO4
Concentration 50ppm ±1ppm
Chemical Name or Material Phosphate (as Phosphorus) Standard
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS Max % 99.98
Assay Percent Notes as P
Traceability to NIST Traceable to NIST
Identification Passes Test
Density 1g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature