missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphate (as Phosphorus) Standard, (1mL = 0.05mg P), Certified, 50ppm ±1ppm, LabChem™
Supplier: LabChem LC186007
Specifications
Phosphate (as Phosphorus) Standard | |
(1mL = 0.05mg P) | |
Poly Bottle | |
99.98,0.02 | |
as P | |
KH2PO4 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
Passes Test | |
Soluble in water |
50ppm ±1ppm | |
7778-77-0,7732-18-5 | |
125 mL | |
Colorless | |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
Certified | |
1g/cm3 | |
Liquid |
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
516951 | |
potassium dihydrogen phosphate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature