Learn More
Phosgene iminium chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 172211000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phosgene iminium chloride | |
33842-02-3 | |
Yellow | |
97.5% min. (Argentometry) | |
C3H6Cl3N | |
MFCD00011870 | |
04,135; 05,195; 06,170; 08,156 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
dichloromethylidene(dimethyl)azanium;chloride | |
2723945 | |
98% |
98% | |
181°C to 185°C | |
Authentic | |
Glass bottle | |
Cl2C=N(CH3)2Cl | |
100 g | |
15, 10174 | |
Solubility in water: hydrolysis | |
[Cl-].CN(C)[C+](Cl)Cl | |
162.44 | |
162.45 | |
Crystalline Powder |
Chemical Identifiers
33842-02-3 | |
162.44 | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
2723945 |
C3H6Cl3N | |
MFCD00011870 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
[Cl-].CN(C)[C+](Cl)Cl |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 251-695-4
RUO – Research Use Only