Please login to your online account to display your discounted pricing

Phorbol 12-myristate 13-acetate, Tocris

Protein kinase C activator

$95.70 - $316.20

Chemical Identifiers

Chemical Name or Material Phorbol 12-myristate 13-acetate
Assay Percent Range >99%
Molecular Formula C36H56O8
Formula Weight 616.83
Synonym (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a- (Acetyloxy)-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1H-cyclopropa[3,4]benz[1,2-e]azulen-9-yl tetradecanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

12011

 
r & d systems
RDS12011
1mg Each for $95.70

12015

 
r & d systems
RDS12015
5mg Each for $316.20
Description & Specifications

Specifications

Formula Weight 616.83
Quantity 1mg
CAS 16561-29-8
Chemical Name or Material Phorbol 12-myristate 13-acetate
Molecular Formula C36H56O8
Assay Percent Range >99%
Synonym (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a- (Acetyloxy)-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1H-cyclopropa[3,4]benz[1,2-e]azulen-9-yl tetradecanoate
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C