Please login to your online account to display your discounted pricing

Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film), Fisher BioReagents

$180.10 - $180.10

Chemical Identifiers

Chemical Name or Material Phorbol 12-Myristate 13-Acetate
CAS 16561-29-8
Physical Form Solid
Assay Percent Range ≥99.0 %
Molecular Formula C36H56O8
Formula Weight 616.8
Synonym PMA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP685-1

  SDS
fisher scientific company
FLBP6851
Glass Ampule 1mg Each for $180.10
Description & Specifications

Specifications

Formula Weight 616.8
Melting Point 72°C
Quantity 1mg
Physical Form Solid
Molecular Formula C36H56O8
Color Off-white
Chemical Name or Material Phorbol 12-Myristate 13-Acetate
Packaging Glass Ampule
Purity 99.0% min.
Assay Percent Range ≥99.0 %
CAS 16561-29-8
ChemAlert Storage Symbol Gray
Synonym PMA
Recommended Storage -20°C
Health Hazard 1 Signal Word:WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health: 1
Flammability:

PMA is an activator of Protein Kinase C; it is a skin irritant and mouse skin tumor promoter.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Signal Word:WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health: 1
Flammability:

-20°C