Learn More
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131041000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phloroglucinol dihydrate | |
218°C to 221°C | |
Authentic | |
Glass Bottle | |
C6H3(OH)3·2H2O | |
100 g | |
15, 7439 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and pyridine | |
C1=C(C=C(C=C1O)O)O.O.O | |
162.14 | |
162.14 | |
Crystalline Powder |
6099-90-7 | |
Beige to White | |
98.5% min. (HPLC) | |
C6H6O3·2H2O | |
MFCD00149090 | |
06, 1092 | |
phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate | |
MPYXTIHPALVENR-UHFFFAOYSA-N | |
benzene-1,3,5-triol;dihydrate | |
80196 | |
99% |
Chemical Identifiers
6099-90-7 | |
162.14 | |
MPYXTIHPALVENR-UHFFFAOYSA-N | |
80196 | |
C1=C(C=C(C=C1O)O)O.O.O |
C6H6O3·2H2O | |
MFCD00149090 | |
phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate | |
benzene-1,3,5-triol;dihydrate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only